Learn More
ATCC Quantitative genomic DNA, Shigella flexneri strain 24570 - 100 μL
Supplier:  ATCC 29903DQ
Encompass
Quantitative genomic DNA prepared from Shigella flexneri strain 24570 supplied as a validated, quantified standard for molecular assay development, verification, validation, and performance monitoring.
  • Genomic DNA prepared from Shigella flexneri strain 24570.
  • Supplied as 100 μL of quantified DNA suitable for qPCR standard curves and assay calibration.
  • Intended for assay development, verification, validation, and monitoring of molecular assays.
  • Store at -20°C or colder to preserve integrity.
  • For research use only; not for human or animal therapeutic or diagnostic use.
Catalog No. NC3802052