Learn More
ATCC Quantitative Genomic DNA from Ureaplasma urealyticum T-strain 960 - qPCR standard
Supplier: ATCC 27618DQ
Quantitative genomic DNA from Ureaplasma urealyticum T-strain 960 is a frozen molecular standard intended for assay development, verification, validation, and routine monitoring of molecular-based assays. The quantitative format enables generation of qPCR standard curves to determine bacterial load and support assay performance evaluation.
- Organism: Ureaplasma urealyticum T-strain 960.
- Format: Frozen quantitative genomic DNA.
- Concentration: ≥1 x 10^5 copies/μL.
- Intended use: For laboratory research use only; not for therapeutic, diagnostic, or consumption purposes.
- Storage: Store at -20°C or colder.
- Handling: Thaw at room temperature, place immediately on ice, briefly centrifuge before opening, and aliquot to avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.