Learn More
ATCC Quantitative Genomic RNA from Influenza A Virus (H3N2) - strain A/Baltimore/JH-0440/2022, 100 μL (VR-3401D)
Supplier: ATCC VR3401D
This quantitative genomic RNA preparation is intended for assay development, verification, and qPCR standard curve generation to determine viral load.
- Material type: genomic RNA.
- Organism/strain: Influenza A virus (H3N2) - A/Baltimore/JH-0440/2022.
- Packaging/volume: 100 μL.
- Storage conditions: -70°C or colder.
- Product format: frozen.
- Biosafety level: BSL 1.
- Verification method: whole-genome sequencing.
- Applications: assay development, verification and validation, infectious and respiratory disease research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.