Learn More
ATCC Quantitative Synthetic Influenza A Virus (H3N2) RNA standard, 100 μL (ATCC VR-3387SD)
Supplier:  ATCC VR3387SD
Encompass
Quantitative synthetic Influenza A (H3N2) RNA intended as a quantitative molecular standard for assay development, verification, validation, and qPCR standard curve generation. The material comprises two synthetic constructs that together cover key gene regions and is synthetically derived, non-infectious, and non-replicative.
  • Contains two synthetic constructs that together cover HA, NP, NA, M1/M2, and NEP/NS1.
  • Designed for qPCR standard curve generation and molecular assay validation.
  • Synthetically derived and non-infectious; cannot replicate.
  • Supplied frozen; store at -70°C or colder.
  • Handled under biosafety level 1 conditions.
  • Unit size 100 μL for use in assay development and verification.
Catalog No. NC3886072