Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Block Upgrade Kit (with Accufill™ System)
GSA_VA

Catalog No. 4471067
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 block
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-710-67 1 block
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-710-67 Supplier Applied Biosystems™ Supplier No. 4471067

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

missing translation for 'validMainframeMsg'

Accelerate genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.

Plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the AccuFill™. Easily load 4 arrays into the QuantStudio 12K Flex instrument to achieve over 12,000 data points.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format OpenArray™ Plate
Content And Storage Store at Room Temperature
For Use With (Equipment) QuantStudio™
Product Line QuantStudio
Product Type Block Upgrade Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 block
Plate Compatibility OpenArray™ Plate
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.