Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio 3 0.1 mL and SYBR Master Mix Starter Package PROMO

Catalog No. A38623PM
Encompass_Preferred
Change view
Click to view available options
Capacity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
A38623PM 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A38623PM Supplier Applied Biosystems™ Supplier No. A38623PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

QuantStudio 3 and SYBR Master Mix Starter Package is a complete qPCR workflow bundle featuring QuantStudio 3 system with laptop, SuperScript IV VILO Master Mix with ezDNase, PowerUp SYBR Master Mix, 96-well fast plates, optical adhesive covers, and SmartStart setup.

Streamline your real-time PCR workflow with this comprehensive, ready-to-run solution designed for reliable, high-performance gene expression analysis. The QuantStudio™ 3 0.1 mL qPCR System package combines advanced instrumentation, optimized reagents, and essential consumables—paired with a dedicated laptop and SmartStart setup—to enable fast implementation and consistent results from day one.

Ideal for research laboratories seeking efficiency, sensitivity, and reproducibility, this bundle eliminates the need to source individual components while ensuring full compatibility across the entire qPCR workflow.

What's Included

  • QuantStudio™ 3 0.1 mL Real-Time PCR System
  • Pre-configured laptop for instrument control and data analysis
  • SmartStart installation and onboarding support for QuantStudio™ 3
  • SuperScript™ IV VILO™ Master Mix with ezDNase™
  • PowerUp™ SYBR™ Green Master Mix (5 x 5 mL)
  • 96-well fast thermal cycling plates
  • MicroAmp™ optical adhesive covers

Features

  • Complete, integrated workflow: Includes instrument, software-ready laptop, reagents, and consumables in one convenient package.
  • High-performance qPCR system: QuantStudio™ 3 delivers precise thermal control, sensitive fluorescence detection, and intuitive software for dependable data collection and analysis.
  • Fast implementation: SmartStart support helps accelerate installation, setup, and onboarding so labs can begin running experiments quickly.
  • Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ supports efficient cDNA synthesis while reducing genomic DNA carryover.
  • Robust SYBR Green chemistry: PowerUp™ SYBR™ Green Master Mix supports strong amplification performance, specificity, and reproducibility.
  • Efficient plate-based workflow: 96-well fast thermal cycling plates support rapid cycling and streamlined sample processing.
  • Reliable sealing for consistent results: Optical adhesive covers help minimize evaporation and support accurate fluorescence detection.

Benefits

  • Accelerate lab readiness with a bundled solution that reduces setup time and simplifies implementation.
  • Reduce purchasing complexity by sourcing a compatible qPCR workflow under a single SKU.
  • Improve confidence in results with trusted instrumentation, optimized chemistries, and workflow-ready consumables.
  • Support reproducibility through integrated components designed to work together across the qPCR process.
  • Increase productivity with a complete package suitable for gene expression analysis and routine qPCR applications.

This all-in-one qPCR workflow bundle is designed to help laboratories simplify procurement, reduce implementation time, and generate high-quality real-time PCR results with confidence.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Non-interchangeable
Capacity 1 x 96-well Plate
Voltage 100/240 V
Product Type Real Time PCR Instrument
No. of Programs 1000
Block Configurations 96-well FAST
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.