Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio 3 0.1 mL and TaqMan Master Mix Starter Package PROMO

Catalog No. A38706PM
Encompass_Preferred
Change view
Click to view available options
Capacity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
A38706PM 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A38706PM Supplier Applied Biosystems™ Supplier No. A38706PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complete qPCR workflow bundle featuring QuantStudio 3 0.1 mL system with laptop, SuperScript IV VILO Master Mix with ezDNase, TaqMan Fast Advanced Master Mix, 96-well fast plates, optical adhesive covers, and SmartStart setup for fast, precise gene expression analysis.

Enable fast, precise, and reproducible real-time PCR with this comprehensive QuantStudio™ 3 0.1 mL qPCR workflow bundle. Designed to support high-performance gene expression analysis, this package combines advanced instrumentation, optimized reagents, and essential consumables—along with a dedicated laptop and SmartStart onboarding support—to streamline implementation and accelerate results.

This all-in-one solution is ideal for laboratories seeking a complete, compatible workflow for TaqMan™-based assays, reducing the complexity of sourcing individual components while ensuring consistent performance across every step of the qPCR process.

What's Included

  • QuantStudio™ 3 0.1 mL Real-Time PCR System
  • Pre-configured laptop for instrument control and data analysis
  • SmartStart installation and onboarding support for QuantStudio™ 3
  • SuperScript™ IV VILO™ Master Mix with ezDNase™
  • TaqMan™ Fast Advanced Master Mix
  • 96-well fast thermal cycling plates
  • MicroAmp™ optical adhesive covers

Features

  • Complete qPCR workflow solution: Includes instrument, laptop, reagents, and consumables in a single bundle to simplify procurement and ensure compatibility.
  • High-performance real-time PCR system: The QuantStudio™ 3 system delivers precise thermal cycling, sensitive fluorescence detection, and intuitive software for reliable data acquisition and analysis.
  • Fast cycling capability: 0.1 mL block format and fast thermal cycling plates enable rapid run times without compromising data quality.
  • Accelerated onboarding with SmartStart: Guided installation and setup support help users quickly begin generating results with confidence.
  • Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ supports efficient cDNA synthesis and helps reduce genomic DNA contamination.
  • High-specificity probe-based chemistry: TaqMan™ Fast Advanced Master Mix enables sensitive, precise quantification with reduced background and high reproducibility.
  • Efficient plate-based workflow: 96-well fast plates support streamlined sample processing for routine and higher-throughput applications.
  • Reliable sealing for consistent results: Optical adhesive covers minimize evaporation and ensure uniform fluorescence detection across wells.

Benefits

  • Accelerate time to data with a fully integrated system designed for fast setup and rapid thermal cycling.
  • Simplify purchasing and setup by consolidating all essential qPCR components into one convenient SKU.
  • Enhance data accuracy and specificity using TaqMan™ probe-based chemistry for precise gene expression analysis.
  • Improve reproducibility with optimized reagents and instrumentation designed to work seamlessly together.
  • Increase laboratory efficiency by reducing workflow complexity and minimizing preparation steps.
  • Support a range of applications including gene expression analysis, assay validation, and routine real-time PCR experiments.

This QuantStudio™ 3 0.1 mL qPCR workflow bundle with TaqMan™ chemistry provides a reliable, high-performance solution for laboratories looking to streamline operations and generate consistent, high-quality real-time PCR results.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Non-interchangeable
Capacity 1 x 96-well Plate
Voltage 100/240 V
Product Type Real Time PCR Instrument
No. of Programs 1000
Block Configurations 96-well FAST
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.