Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio 3 0.2 mL and SYBR Master Mix Starter Package PROMO

Catalog No. A39941PM
Encompass_Preferred
Change view
Click to view available options
Capacity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
A39941PM 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39941PM Supplier Applied Biosystems™ Supplier No. A39941PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

Complete qPCR workflow bundle featuring the QuantStudio 3 0.2 mL system with laptop, SuperScript IV VILO Master Mix with ezDNase, PowerUp SYBR Master Mix, MicroAmp 96-well plates, optical adhesive covers, and SmartStart setup for fast, reliable gene expression analysis.

Enable efficient and reliable real-time PCR with this comprehensive, ready-to-use workflow solution. The QuantStudio™ 3 0.2 mL qPCR System bundle combines advanced instrumentation, optimized reagents, and essential consumables—along with a dedicated laptop and SmartStart onboarding support—to streamline setup and accelerate results from day one.

Designed for research laboratories focused on gene expression analysis and routine qPCR applications, this all-in-one package simplifies procurement while ensuring compatibility across every step of the workflow.

What's Included

  • QuantStudio™ 3 0.2 mL Real-Time PCR System
  • Pre-configured laptop for instrument control and data analysis
  • SmartStart installation and onboarding support for QuantStudio™ 3
  • SuperScript™ IV VILO™ Master Mix with ezDNase™
  • PowerUp™ SYBR™ Green Master Mix (5 x 5 mL)
  • MicroAmp™ 96-well reaction plates
  • MicroAmp™ optical adhesive covers

Features

  • Complete qPCR workflow solution: Includes instrument, software-ready laptop, reagents, and consumables in a single, convenient bundle to reduce sourcing complexity.
  • Flexible 0.2 mL block format: Supports standard reaction volumes and compatibility with widely used plate formats for versatile lab workflows.
  • High-performance real-time PCR system: The QuantStudio™ 3 system delivers precise thermal cycling, sensitive fluorescence detection, and intuitive software for consistent, high-quality results.
  • Rapid setup with SmartStart support: Guided installation and onboarding help users quickly implement the system and begin generating data with confidence.
  • Optimized reverse transcription: SuperScript™ IV VILO™ Master Mix with ezDNase™ enables efficient cDNA synthesis while helping reduce genomic DNA contamination.
  • Reliable SYBR Green chemistry: PowerUp™ SYBR™ Green Master Mix supports robust amplification performance, high specificity, and reproducibility across experiments.
  • Efficient plate-based workflow: MicroAmp™ 96-well plates support streamlined sample processing and consistent thermal performance.
  • Secure sealing for data integrity: Optical adhesive covers minimize evaporation and ensure reliable fluorescence detection.

Benefits

  • Accelerate time to results with a fully integrated system that reduces setup time and enables immediate workflow implementation.
  • Simplify procurement by consolidating instrumentation, reagents, and consumables into a single SKU.
  • Ensure workflow compatibility with components designed and validated to work seamlessly together.
  • Enhance data reliability through precise instrumentation and optimized reagent performance.
  • Support reproducibility across gene expression and routine qPCR applications.
  • Improve lab efficiency with a complete solution that reduces coordination across multiple suppliers.

This all-in-one QuantStudio™ 3 0.2 mL qPCR workflow bundle is designed to help laboratories streamline operations, reduce implementation time, and achieve consistent, high-quality real-time PCR results.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Non-interchangeable
Capacity 1 x 96-well Plate
Voltage 100/240 V
Product Type Real Time PCR Instrument
No. of Programs 1000
Block Configurations 96-well
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.