Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 7 Flex Real-Time PCR System, 384-well, laptop
GSA_VA

Catalog No. 4485695
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-856-95 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-856-95 Supplier Applied Biosystems™ Supplier No. 4485695

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

missing translation for 'validMainframeMsg'

Includes

Laptop Computer

Advances your research further by enabling a broad range of real-time PCR-based applications through its multiplexing capabilities and interchangeable block formats.

With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.

Features of the QuantStudio™ 7 Flex Real-Time PCR System include:
  • Increased application versatility through the accommodation of 4 different block types
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, automation capabilities, and effortless block exchange without the need for any tools

Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

The QuantStudio™ 7 Flex system accommodates the interchange of a 96-well, 96-well Fast, 384-well, or TaqMan™ array card block. The system is delivered with one block type (a 384-well block in this case); other block types may be purchased separately. In addition, a Twister™ Robot Automation Accessory is available for this system to support high throughput workflows. For a more value-priced system that does not support TaqMan™ array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio™ 6 Flex system. And for the ultimate in flexibility, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan™ OpenArray™ plates.

Accomplish More, Faster:
Run hundreds of real-time PCR reactions effortlessly using TaqMan™ Array 384-well microfluidic cards. And when integrated with the Twister™ Robot Automation Accessory (available separately), the QuantStudio™ 7 Flex system maximizes your throughput capabilities.

High Application Versatility:
Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, High Resolution Melt (HRM), and Protein Thermal Shift™ experiments.

Results You Can Trust:
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 6 decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.

Sensitive Data Analysis:
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

For Research Use Only. Not for use in diagnostic procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Interchangeable
Format 384-well Plate
Content And Storage Laptop Computer
For Use With (Equipment) QuantStudio™
Product Line QuantStudio
Display Type Touchscreen
Quantity 1 system
When using the QuantStudio 6 or QuantStudio 7 Software version 1.0, I get the error message "Unable to start the run" when starting a new run. What can I do?

We recommend that you send the run file to the QuantStudio 6 or QuantStudio 7 Real-Time PCR instrument and start the run from the instrument touchscreen. Additionally, you can upgrade to v1.3 or above of the QuantStudio Real-Time PCR software and you will not have this error message show up.

How do I activate or renew my software license in the QuantStudio 6 or QuantStudio 7 Real-Time PCR System?

Please follow the instructions found in the Software Licensing Quick Reference Guide at https://fnoclient.gss.tf/quick-start-guide to activate or renew a license for the QuantStudio 6 or QuantStudio 7 Real-Time PCR System. If you have a license key associated with a computer that is no longer in use, please follow the instructions to "Transfer a license to a different user or computer" in the above guide.

What should I do if the software will not connect to the QuantStudio 6 or QuantStudio 7 Real-Time PCR System?

After launching the QuantStudio software, go to the Maintenance Manager. Check if the QuantStudio system icon is present in the “On the Network” window. If not, click on the ‘Refresh' button to the left of ‘Remove from My Instruments'. If you still do not see the QuantStudio system icon for the following reasons:

- There is an icon but you cannot add it into the "My Instruments" area
- There is a red “X” on the instrument
- A message says "Disconnected" at the bottom of the window with a red “X” in it
then you will need to add the Instrument into the ‘My Instruments' area. Click on the wrench icon in the upper right-hand corner of the icon as pictured below. You may get a message that says the firmware is older than the one you're trying to run. If appropriate, choose to download the new firmware and let it progress to finish. Do not turn off the instrument during this update. After the firmware update is completed, restart the instrument and check for the QuantStudio system icon in ‘On the Network' area.

What can I do when I get an error message related to "...JVM..."?

This error can occur due to allocation of the computer's virtual memory. If you see this message, try the following:

1. Click ‘OK' to the error, close any unnecessary programs, and re-launch the software (you may have to do this several times).
2. Using the Task Manager (Control+Alt+Delete), look for a process called ‘javaw.exe'. Select this line and end the process. Then re-launch the software.

On the QuantStudio 6 and 7 Flex instruments, what does the error “Calibration file already exists” mean?

The calibration files are stored on the QuantStudio 6/7 Flex instrument itself; therefore, when you need to perform a new calibration, you will have to delete the old calibration files. From the instrument console, go to the QuantStudio 6/7 Flex instrument touchscreen and select ‘Collect Results'. You should then see a list of files stored on the instrument. Manually select a calibration file such as “Fast-96-cal”, and press the ‘Delete' button to remove the old calibration file. Occasionally, users cannot delete the file off the instrument and may encounter this error: “Some experiments did not collect results therefore cannot be deleted.”. In these situations, launch the software, go to ‘maintenance', manage files, and download the calibration file. The status of the experiment should be displayed as ‘complete'. Next, delete the old calibration file and rerun the calibration.

Is it possible to see the traces in an allelic discrimination experiment on the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

The ‘reveal traces' feature for genotyping runs allows you to trace the clusters throughout the PCR process. For some assays you may find better cluster discrimination at, say, cycle 40, as opposed to cycle 50. To use this feature, the amplification data needs to have been collected for the run.

- Go to ‘Analysis Settings'
- Under the default ‘Call Settings' tab, Choose ‘Analyze Real-Time Rn Data'
- Click ‘Apply Analysis Settings'
- Under the Allelic Discrimination Plot, check the box next to ‘Reveal Traces'. You can then move the slide bar to see how the data changes with the cycles number.
In the example below, the image on the left shows the data at a full 40 cycles. The image on the right is the same data after revealing traces (grey lines). The clusters can be traced back by moving the cycle bar. Notice that one point is no longer called at the earlier cycle set point.

How do I transfer a QuantStudio software license to another computer?

Follow the directions in this blog post (http://solid.community.appliedbiosystems.com/community/real-time_pcr/blog/2013/03/26/transferring-a-quantstudio-12k-flex-software-license-to-another-computer) to transfer your license to another computer. (Please note that the instructions here still apply, although they are written for the QuantStudio 12K Flex Real-Time PCR System.)

Where are my files saved on the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

Run files will be saved to both the instrument and the connected computer. On the connected computer, files will be saved to the default data folder, unless you change it.

To find or change the default folder, go to then Tools then Preferences then Defaults. Here you will see a Data Folder and an Import Folder. The default location is shown. If you want files to be saved to (or open from) a different location, click ‘Browse', and choose the new folder.

Can I collect data at two separate steps for analysis in the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

Yes, the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems can collect data at multiple steps in the amplification stage. You will need to turn on the data collection for every step of interest. In the analysis, only one set of data can be displayed at a time. To change the data set used for analysis, go to Analysis Settings then Ct Settings and change the drop down under ‘Data Step Selection'.

What volumes can be used in my plate on the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR System?

The following volumes are supported for each instrument block:

-96-well standard (10-100 µL reactions)
-96-well Fast (10-30 µL reactions)
-384-well (5-20 µL reactions)
-TaqMan Array Micro Fluidic Cards (approximately 1 µL reactions) - for QuantStudio 7 Flex Real-Time PCR System only

How many files can the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems hold?

Each instrument can hold up to 100 experiment files.

How can I add a custom dye to the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

You will need an oligo sequence with the custom dye but without the quencher molecule. Set up a plate of dye dilutions such as seen in the “Custom Dye Calibration” section of this link:
https://tools.thermofisher.com/content/sfs/manuals/4474347B.pdf
1.Set up a dummy run using the ‘Standard Curve' option. Alter the thermal profile so that it simply ramps to 60 degrees C with a 2 min hold. Ensure that the filters of interest are selected.
2.When the run is complete, export and examine the raw data. Select the concentration to use by finding the dilution that will give you an acceptable signal in the following ranges:
- For a 384-well plate: between 400,000 and 1,200,000
- For a 96-well plate: between 1,400,000 and 4,300,000
3.Create a full plate of the dye using the selected concentration and run the custom dye calibration as normal, using 20 µL

How do I set up a touch-down PCR on the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

In a touchdown PCR experiment, you will change either the temperature or the time of a particular PCR step with every cycle. Most commonly, the annealing temperature is adjusted throughout the experiment, such that the specificity is higher in the early cycles and the efficiency in the later cycles.

1. Go to File then New Experiment then Advanced Setup. Fill out the relevant options as you normally would.
2. Go to the Run Method under the ‘Setup' section and you should see the Graphical View of your thermal profile. Check the box next to ‘Enable AutoDelta'. You should see some grey triangles appear next to the Temperature and Time at every step in the Cycling Stage. (Note: If you want to start the changes at a later cycle, set this here under ‘Starting Cycles'.)
3. A new window called ‘AutoDelta Settings' will open up. Select the appropriate options. In this example we are decreasing the temperature by 0.4 degrees C per cycle, so choose (“-“) and (0.40). Click ‘Save Setting'. You will then see a green triangle show up next to the parameter you changed, in this case next to the 72 degrees C step. Your new method has now been applied.

What is the difference between the QuantStudio 6 and QuantStudio 7 Flex Real-Time PCR Systems?

Please see the comparison table in this brochure (https://www.thermofisher.com/content/dam/LifeTech/migration/files/pcr/pdfs.par.56935.file.tmp/co06402%20quantstudio%20family%20brchr-final-web.pdf) for details on the differences between the two instruments.

What are the uniformity and normalization calibrations for?

The uniformity calibration generates data that allows the software to compensate for the physical effects of the QuantStudio system filters using the ROI (Region of Interest) plate. The normalization calibration generates factors that the software uses when comparing data from multiple QuantStudio instruments within a study. There are 2 normalization plates provided in the calibration kit, one containing FAM/ROX dye and the other with VIC/ROX dye.

When do I need to calibrate the QuantStudio 6 or QuantStudio 7 Flex Real-Time PCR System?

The maintenance schedule can be found in the QuantStudio 12K Flex Maintenance and Administration Guide (Chapter 2). In short, the background calibration should be performed monthly, while all other calibrations should be performed every 6 months.

How do I renew my software license?

User Bulletin v1.2.1 has specific directions for how to renew your software license. Please search the term '4474347' on our website. The renewal directions apply to the ViiA 7, QuantStudio 6, QuantStudio 7, and QuantStudio 12K Flex systems.

How do I know if my software license has expired?

You can check your license status by opening the instrument software and going to Tools --> License Central. You will see the license status and the expiration date.

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?

The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:

  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.


If the problem persists after reseating the block, please contact our Technical Support team for further assistance.

How do I download the Pathogen Interpretive Software?

Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.

With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?

Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)

Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip

Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.

What is the instrument software configuration required to use the TaqPath Monkeypox/Orthopox Virus DNA Kit?

To obtain the correct analysis software or firmware version for your real-time PCR instrument, go to thermofisher.com/validatedfirmware, then select your instrument in the Real-Time PCR section.

Which instruments are authorized under the EUA for use with the TaqPath Monkeypox/Orthopox Virus DNA Kit?

Here are the instruments that can be used with the TaqPath Monkeypox/Orthopox Virus DNA Kit:
- Applied Biosystems QuantStudio 5 Real-Time PCR System (96-well, 0.1 mL block)
- Applied Biosystems QuantStudio 5 Dx Real-Time PCR System (96-well, 0.2 mL block)
- Applied Biosystems QuantStudio 7 Flex Real-Time PCR System (384-well block)

I would like to use the TaqPath Monkeypox/Orthopox Virus DNA Kit. Which extraction kit should I use to extract DNA from the sample?

Performance was established using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) in combination with the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head and the KingFisher Flex 96 Deep-Well Heating Block (MVP_2Wash_400_Flex program). Other purification methods may lead to deviations in the expected result.

Which specimen types are compatible with the TaqPath Monkeypox/Orthopox Virus DNA Kit?

The TaqPath Monkeypox/Orthopox Virus DNA Kit protocol is currently authorized for use with lesion swab specimens in universal transport media (UTM) or viral transport media (VTM) from individuals suspected of monkeypox infection by their healthcare provider.

What instruments, consumables, and reagents do I need for the TaqPath Monkeypox/Orthopox Virus DNA Kit? Where can I find the full list of EUA products and protocols?

Go to www.thermofisher.com/monkeypox and download the Instructions for Use (IFU) to see the workflow outlining the instruments, consumables, and reagents required to run Monkeypox detection experiments.

What are the reporter dyes and targets for the TaqPath Monkeypox/Orthopox Virus DNA Kit?

The reporter dyes used are as follows:
- Monkeypox virus: FAM dye
- Orthopoxvirus: VIC dye
- RNase P: JUN dye

The TaqPath Monkeypox/Orthopox Virus DNA Kit probes contain QSY and MGB quenchers, which do not fluoresce. Select None for Quencher in the instrument set up procedure.

What are the components of the TaqPath Monkeypox/Orthopox Virus DNA Kit? Is the master mix included, or does it need to be purchased separately? Are there any other required materials that are not supplied?

The TaqPath Monkeypox/Orthopox Virus DNA Kit includes the following components:
- TaqPath Monkeypox/Orthopox Virus Multiplex Assay: Multiplexed real-time PCR assay containing primers/probes specific to monkeypox virus, non-variola orthopoxvirus targets, and human RNase P.
- TaqPath Monkeypox/Orthopox Virus DNA Positive Control: Positive DNA control that contains templates specific to monkeypox virus, non-variola orthopoxvirus targets, and human RNase P regions targeted by the assay.

Note:
- The master mix is not included in the kit and needs to be purchased separately: TaqPath BactoPure Microbial Detection Master Mix (No ROX) (Cat. No. A52703, A52704, or A52705)
- TE Buffer (Cat. No. 12090015) and Nuclease-free water (MLS) are required and not supplied with the kit.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Real-Time PCR Software?

If you are analyzing results from the ViiA 7 Real-Time PCR System, QuantStudio 6 Flex Real-Time PCR System, or QuantStudio 7 Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Real-Time PCR Software following the instructions below:
1. Open the data file in the QuantStudio Real-Time PCR Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

How do I calibrate the QuantStudio 6 Flex and QuantStudio 7 Flex Real-Time PCR instruments with Cy5 dye?

If you are running software v1.5.1 and below on the QuantStudio 6 Flex or QuantSudio 7 Flex Real-Time PCR instrument and using a 384-well plate, you may reach out to our Specialty Oligos team at Specialty_Oligos@thermofisher.com to request Cy5 dye calibration oligos. Each tube is provided at 800 µL, 0.2 µM. For a 384-well plate, you will need to order five tubes and aliquot 10 µL/well. Please provide your shipping address to Specialty Oligos and they will provide you with a formal quote.

If you are using a 96-well block, the Cy5 dye plate contained in either 7500 Real Time PCR Systems Spectral Calibration Kit II (Cat. No. 4351151) (Standard, 0.2 mL) or 7500 Fast Real-Time PCR Systems Spectral Calibration Kit II (Cat. No. 4362201 (Fast, 0.1 mL) can be used in place of the Cy5 dye calibration oligos.

What qPCR instruments are compatible with Thermo Fisher Cloud?

All our real-time PCR instruments are compatible with Thermo Fisher Cloud Apps except the 7500 Fast instrument where the eds files alone are compatible with Thermo Fisher Cloud Apps.

With the Applied Biosystems QuantStudio 6 and 7 Flex Real-Time PCR System Software, v1.0, I get the error message "Unable to start the run" when starting a new run. What can I do?

Please uncheck and check the “Data Collection” camera icon in the Run Method, and the “Start Run” button should now be active again.

I would like to calibrate my Applied Biosystems QuantStudio 6 and 7 Flex Real-Time PCR instruments with a custom dye, which buffer should I use?

Please ask your dye manufacturer for recommendations. If there is no specific recommendation available, try TE Buffer (pH 8.0) to start.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.