Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 7 Pro Dx Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, SYBR™ dyes), 384-well, IVD

Catalog No. A52439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52439 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52439 Supplier Applied Biosystems™ Supplier No. A52439
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Enables simultaneous multiple dye calibrations in one plate, significantly reducing effort and time required for dye calibration.

This calibration plate establishes the pure dye spectra and multicomponent values for FAM, VIC, ROX, TAMRA, and SYBR dyes on the QuantStudio 7 Pro Dx 384-well real-time PCR system. It enables simultaneous multiple dye calibrations in one plate, significantly reducing effort and time required for dye calibration. Please refer to the instrument user guide for full instructions on usage.

TRUSTED_SUSTAINABILITY

Specifications

Calibrated Dye FAM™, ROX™, TAMRA™, VIC™, SYBR™ Green
For Use With (Equipment) QuantStudio™ 7 Pro Dx
Detection Method qPCR
Product Type Spectral Calibration Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 Plate
Plate Compatibility 384-well
Calibration Type Spectral Calibration

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.