Learn More
Applied Biosystems™ QuantStudio™ Dx Real-Time PCR Instrument with 96-well fast block
Description
Includes
No Computer
The QuantStudio™ Dx instrument is approved for in vitro diagnostic (IVD) use with the Quidel™ Molecular Direct C. difficile Assay, the first available from a growing menu of assays that are under development. In Test Development Mode, the QuantStudio™ Dx Real-Time PCR Instrument offers easy-to-change blocks that can also accommodate array cards, as well as 96-well standard, 96-well Fast, and 384-well plates, allowing you to select the format that is right for your throughput needs. Built on a legacy of innovation and excellence in real-time PCR, the QuantStudio™ Dx instrument leverages the best of our portfolio of Applied Biosystems™ real-time PCR systems, and refines a platform to specifically meet the needs of the diagnostic laboratory.
The QuantStudio™ Dx Real-Time PCR Instrument's proven performance, security, and reliability - combined with the outstanding sensitivity and specificity of the Quidel™ Molecular Assays - help ensure the right answer is delivered the first time, every time.
Flexibility You Want, Security You Need:
In Test Development Mode, The QuantStudio™ Dx Real-Time PCR Instrument with the 96-well fast Block is designed with easily interchangeable thermal cycling blocks to accommodate your laboratory's varying needs for throughput, format type, and assay type. In addition to the 96-well fast format that was cleared for use in the US with the Quidel Molecular Direct C. difficile Assay, the instrument also accepts 96-well standard, 384-well, and array card formats that can be used in Test Development Mode for assay development.
Efficient Workflows and Assays:
A simplified instrument and assay workflow, as well as intuitive software, make the entire test easy and efficient, from processing to reporting. Plus, it frees up highly trained operators to perform more complex tasks.
QuantStudio™ Dx Software:
- Track reagent and patient information for quality control purposes
- Enter reagent information by barcode and upload patient information from Excel files to minimize data entry errors
- Print calibration and maintenance reports for your instrument records
- Included SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability as part of 21 CRF Part 11 compliance
Available in select countries only.
Specifications
Specifications
| Block Format | Interchangeable |
| Format | 96-well Plate |
| Temperature Range (Metric) | 4 to 100°C |
| Content And Storage | 96-well Fast Block |
| For Use With (Equipment) | QuantStudio™ Dx |
| Thermal Uniformity | ±0.5°C |
| Reaction Speed | Fast |
| Dimensions | 53.3 cm (21 in.) (W) x 63.5 cm (25 in.) (D) x 64.5 cm (25.4 in.) (H) |
| Product Line | QuantStudio |
| Quantity | 1 instrument |
Frequently Asked Questions (FAQs)
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:
- Remove the block from the instrument.
- Reinsert the block into the instrument.
If the problem persists after reseating the block, please contact our Technical Support team for further assistance.
Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.