Learn More
Glass Expansion Quartz Torch, 1.2mm, Shimadzu ICPM-8500
SDP
Supplier:  Glass Expansion 308089986
Encompass_Preferred
Quartz Torch, single-piece, 1.2 mm id injector, for Shimadzu ICPM-8500 ICP-MS (211-44604-1). Suitable for organic applications with volatile solvents but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7045