Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ 4 Starter Packages, with WiFi

Catalog No. p-7235449
Encompass_Preferred
Change view
Click to view available options
Type:
NGS Starter Kit, w/ WiFi
Prot. Broad Range Starter Kit
Quant Starter Kit, w/ WiFi
RNA IQ Starter Kit, with WiFi
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
Q33239 Quant Starter Kit, w/ WiFi
Q33240 NGS Starter Kit, w/ WiFi
Q33241 RNA IQ Starter Kit, with WiFi
A51292 Prot. Broad Range Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Q33239 Supplier Invitrogen™ Supplier No. Q33239
Only null left
Add to Cart
Add to Cart
Request a Quote

Accurately quantify protein, dsDNA, and RNA with the Qubit 4 Protein BR, Quantitation, NGS, and RNA IQ Starter Kits, which feature a fluorescent dye that emits signal only when bound to its target molecule, minimizing the effect of contaminants.

Achieve accurate and reliable results with Qubit 4 Starter Packages, which combine the Qubit 4 Fluorometer with different assay kits for the quantification of nucleic acids or proteins. Qubit reagents bind selectively to DNA, RNA, or protein and only emit a fluorescent signal when bound to the target. This minimizes the effects of contaminants on the result and ensures accurate readings for sensitive samples.

With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result.  Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds.  Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.

Qubit 4 Quantitation Starter Kit

The Qubit 4 With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result. Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds. Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.

Qubit 4 Quantitation Starter Kit (Cat. No. Q33239) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit 4 System Verification Assay Kit, 50 assays (Cat. No. Q33237)
• Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33230)
• Qubit 1X dsDNA BR Assay Kit, 100 assays (Cat. No. Q33265)
• Qubit RNA IQ Assay Kit, 75 assays (Cat. No. Q33221)
• Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
• Qubit RNA BR Assay Kit, 100 assays (Cat. No. Q10210)
• Qubit Protein BR Assay Kit, 100 assays (Cat. No. A50668)

Qubit 4 NGS (Next-Generation Sequencing) Starter Kit
The Qubit NGS Starter Kit (Cat. No. Q33240) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Wi-Fi Dongle (Cat. No. A26774)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)

Qubit Protein BR Starter Kit
The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, and complex protein mixtures. The assay can deliver accurate protein quantification across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods.

The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use with the Qubit 4 Fluorometer. The assay is not designed for use with other Qubit models. The kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then, insert your sample into a Qubit 4 instrument and read the concentration.

The Qubit 4 Protein BR Starter Kit (Cat. No. A51292) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Wi-Fi Dongle (Cat. No. A26774)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit Protein BR Assay Kit, 100 assays (Cat. No. A50668)

Qubit 4 RNA IQ Starter Kit
The Qubit RNA IQ assay provides a fast, simple method to check whether an RNA sample has degraded. The assay utilizes two unique dyes: one that binds to large, intact RNA, and another that selectively binds to small, degraded RNA. Together they enable you to quickly assess the quality and integrity of an RNA sample. To use, simply add your samples to the RNA IQ working solution, then measure on the Qubit 4 Fluorometer.

The Qubit 4 RNA IQ Starter Package (Cat. No. Q33241) includes:
• Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
• Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
• Qubit RNA IQ Assay Kit, 275 assays (Cat. No. Q33222)

TRUSTED_SUSTAINABILITY

Specifications

Sample Volume 1 to 20 μL
Includes Fluorometer, Assay Tubes, Wi-Fi Dongle, Assay Kit
Excitation Wavelength 470 nm (Blue LED), 635 nm (Red LED)
For Use With (Equipment) Qubit Fluorometer
Product Line Qubit
Type Quant Starter Kit, w/ WiFi
For Use With (Application) sdDNA Quantification, RNA Quantification, Protein Quantification
Quantity 1 kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.