Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qubit™ Flex Fluorometer with 3-year Warranty

Catalog No. A44985
Encompass_Preferred
Click to view available options
Includes:
3-year AB Repair Center Warranty
3-year Rapid Exchange Warranty

Experience accurate, specific, and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin with the Qubit Flex Fluorometer. Measure up to eight samples simultaneously using Qubit assay kits for reproducible results in seconds.

The Qubit Flex Fluorometer is designed for highly accurate and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin. This benchtop fluorometer offers the flexibility to directly measure fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results. The fluorometer has a large screen and intuitive user interface, making it easy to select and run your desired assay with results displayed in just a few seconds.

Features of the Qubit Flex Fluorometer include:

  • Improved throughput—measure multiple samples and quantify DNA, RNA, protein, and endotoxin up to 50% faster than single-sample readers
  • Flexibility—measure up to eight samples simultaneously
  • Complete optimized system—works seamlessly with Qubit reagents to generate reliable, sensitive, and specific results
  • Accurate quantification—high levels of accuracy using only 1–20 μL of sample, even with very dilute samples
  • On-board calculators—quickly generates Qubit working solution preparation instructions and determines the molarity of your samples
  • Easy to use—a large 8-inch, state-of-the-art color touchscreen and intuitive user interface results in easy workflow navigation
  • Data export flexibility—data is transferred easily using the included WiFi dongle, a USB drive, or connection to your network drive via LAN or Wi-Fi connection
  • Several languages available—choices include English, French, Spanish, Italian, German, simplified Chinese, and Japanese
  • Verification test—a pre-loaded program is included to verify performance of the instrument
  • Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
  • Three-year warranty—warranty packages include either the

Higher throughput, less variability than single sample readers

The Qubit Flex Fluorometer has the flexibility to measure multiple DNA, RNA, protein, and endotoxin samples up to 50% faster than single-sample readers. In addition to saving time, the ability to measure eight samples simultaneously reduces variability, resulting in highly reproducible data. The fluorometer includes reagent, range, and molarity calculators, further reducing the time to results. The reagent and range calculators help to determine which reagent and what amount of sample to use to effectively quantify the DNA, RNA, or protein. The molarity calculator allows you to calculate the molarity of your samples based on nucleic acid length and the measured concentration.

Enables more accurate results than UV absorbance at low concentrations

The Qubit Flex Fluorometer is far more sensitive than UV absorbance, which measures anything absorbing at 260 nm, including DNA, RNA, protein, free nucleotides, and excess salts. Moreover, UV spectrophotometry is often not sensitive enough to accurately measure low concentrations of DNA and RNA. With the Qubit Flex Fluorometer, your research is enhanced by more accurate measurements because the dyes in the Qubit assay kits fluoresce only when bound to the target molecule (DNA, RNA, protein, or endotoxin) in your sample. This allows you to avoid repeating work due to inaccurate measurements.

Optimized solution consisting of instrument and reagents results in highly reproducible data

The thoughtfully designed Qubit Flex Fluorometer requires sample volumes of only 1–20 μL. The advanced optics and data analysis algorithms were designed and optimized to work in perfect harmony with Qubit reagents, resulting in a seamless solution that generates highly reliable, sensitive, and specific results.

Comprehensive warranty options

The Qubit Flex Fluorometer three-year warranty covers all costs for shipping, parts, and labor for repairs. Additional years of coverage can also be purchased to extend coverage. The Qubit Flex Fluorometer packages are offered with two different types of bundled warranties:

  • Three-year Rapid Exchange (REX) Warranty—replacement shipped out in ~1 business day for labs with critical timelines
  • Three-year AB Repair Center (ABRC) Warranty—instrument repaired and returned in ~3 weeks, ideal for regulated labs required to maintain asset tagging consistency
TRUSTED_SUSTAINABILITY

Specifications

Model Qubit Flex Fluorometer
Content And Storage

•Qubit™ Flex Fluorometer
•Qubit™ Flex power cord (shipped separately)
•USB drive (FAT32, 4 GB)
•Qubit™ Flex LAN cable
•Qubit™ Flex Fluorometer Quick Reference Card (QRC)
•Certificate of Conformity (COC)
•Qubit™ screen cleaning cloth
•Wi-Fi Dongle
•Qubit Flex SmartStart Digital Educational Experience
•3-year Rapid Exchange (REX) Warranty or 3-year AB Repair Center (ABRC) Warranty

Includes 3-year AB Repair Center Warranty
Excitation Wavelength 460 to 480 nm (Blue LED), 620 to 640 nm (Red LED)
For Use With (Equipment) Qubit Flex Fluorometer
Product Line Qubit
Type Benchtop Fluorometer
Wavelength Range 400 to 750 nm
Emission Wavelength Green (513-563 nm), Far-red (671-693 nm)
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.