Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits

Catalog No. 03254153
Change view
Click to view available options
Sample Type:
All Food Types
Fruits and Vegetables with Fats and Waxes
General Fruits and Vegetables
Other Foods
Pigmented Fruits and Vegetables
Tube Size:
2 mL
15 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Tube Size
03-254-153 Pigmented Fruits and Vegetables 2 mL
03-254-164 All Food Types 15 mL
03-254-150 Fruits and Vegetables with Fats and Waxes 15 mL
03-254-146 General Fruits and Vegetables 15 mL
03-254-162 Other Foods 15 mL
03-254-163 All Food Types 2 mL
03-254-149 Fruits and Vegetables with Fats and Waxes 2 mL
03-254-145 General Fruits and Vegetables 2 mL
03-254-161 Other Foods 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 03-254-153 Supplier Thermo Scientific™ Supplier No. S22PAOACKIT
Add to Cart
Add to Cart

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Clean-up Kits

These kits use dispersive solid phase extraction (dSPE) to purify supernatants obtained during extraction. Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, fruits and vegetables containing fats and waxes, and all other food types.

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY

Specifications

Description QuEChERS AOAC Clean-up Kit; 50 mg PSA, 50 mg GCB, 150 mg MgSO4 prefilled in 2 mL Tubes; 100/Pk.
Type QuEChERS Clean-up Kit
Includes Salts in Prefilled Tubes
Method QuEChERS sample preparation for AOAC 2007.01 Method
Salt Composition 50 mg PSA, 50 mg GCB, 150 mg MgSO4
Sample Type Pigmented Fruits and Vegetables
Tube Size 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.