Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits

Catalog No. 03254139
Change view
Click to view available options
Includes:
Salts in Pouches
Salts in Pouches, Tubes
Salts in Pouches, Tubes, Ceramic Homogenisers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03-254-139 Salts in Pouches, Tubes
03-254-138 Salts in Pouches, Tubes, Ceramic Homogenisers
03-254-140 Salts in Pouches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-254-139 Supplier Thermo Scientific™ Supplier No. S110ENKIT
Add to Cart
Add to Cart

Use Thermo Scientific™ QuEChERS Extraction Kits for the EN 15662 Method to analyze complex sample matrices for residues, toxins, and other contaminants.

Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Three different extraction kit formats are available for 10 g samples.

  • Extraction Salts in Pouches, no Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

What is the purpose of the chemicals used in the EN 15662 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
  • NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
  • Trisodium Citrate: buffers the sample to stabilize pH
  • Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY

Specifications

Description QuEChERS EN 15662 Method Extraction Kit for 10 g sample; 4 g Anhydrous MgSO4, 0.5 g Disodium Citrate, 1 g Trisodium Citrate, 1 g NaCl in Pouches; Empty 50 mL Tubes; 50/Pk.
Type QuEChERS Extraction Kit
Includes Salts in Pouches, Tubes
Method QuEChERS sample preparation for EN 15662 Method
Salt Composition 4 g Anhydrous MgSO4, 0.5 g Disodium Citrate, 1 g Trisodium Citrate, 1 g NaCl
Sample Size 10 g sample
Tube Size 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.