Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15200310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15200310 10 g
ICN15200325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15200310 Supplier MP Biomedicals, Inc Supplier No. 0215200310
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6151-25-3
Molecular Formula C15H14O9
Molecular Weight (g/mol) 338.27
MDL Number MFCD00149487
InChI Key GMGIWEZSKCNYSW-UHFFFAOYSA-N
Synonym quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate
PubChem CID 5284452
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
SMILES O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Specifications

Color Green-Yellow
Quantity 10 g
Formula Weight 338.26
Percent Purity 0.9985
Physical Form Powder
Chemical Name or Material Quercetin dihydrate, Molecular Biology
EINECSNumber 204-187-1
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.