Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Quetiapine hemifumarate

Catalog No. AC462400250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462400250 25 g
AC462400050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462400250 Supplier Thermo Scientific Chemicals Supplier No. 462400250

missing translation for 'validMainframeMsg'

CAS: 111974-72-2 | C29H33N3O10S | 615.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111974-72-2
Molecular Formula C29H33N3O10S
Molecular Weight (g/mol) 615.65
MDL Number MFCD03423782
InChI Key JLWSQVHJLLYDPX-LVEZLNDCSA-N
IUPAC Name bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 172.0°C to 176.0°C
Color Clear Colorless to Light Yellow
Quantity 25 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in dimethylformamide, slightly soluble in methanol,very slightly soluble, in ether, insoluble in ether and acetonitrile
Formula Weight 441.54
Chemical Name or Material Quetiapine hemifumarate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

Causes damage to organs.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

If eye irritation persists: Get medical advice/attention.

IF exposed or concerned: Get medical attention/advice.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.