accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONNNSOHOOHOOHOOHO(E)(E)

CAS RN 111974-72-2

IUPAC Name: bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
Synonyms: bis(fumaric acid); quetiapine
Molecular Weight (g/mol): 615.65
Molecular Formula: C29H33N3O10S
InChi Key: JLWSQVHJLLYDPX-LVEZLNDCSA-N
SMILES: OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)