Description
- Quick exchange of Series 40 (RC and QR) chambers or 35 mm glass bottom dish
- Includes adapters for Willco Wells, Corning, Falcon, WPI, and MatTek dishes
- Open design for easy access to chamber or dish
- Compatible with Series 20 stage adapters
- Removable perfusion and suction tube holders
- Resistive heating elements for temperature control
- Magnetic stainless steel shelf permits use of magnetic holders
Specifications
Specifications
| For Use With (Equipment) | RC-40/QR-40 Series Chambers |