Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Quick Fluorescent Loading Buffer for Native-PAGE, 3X

Catalog No. ICN08W00014 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN08W00014 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN08W00014 Supplier MP Biomedicals™ Supplier No. 08W00014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Quick Fluorescent Loading Buffer pre-stains a protein sample for Native-PAGE.

Protein samples are stained fluorescently during sample treatment prior to electrophoresis. Protein bands in a Native-PAGE gel or on a membrane after transfer can be visualized and analyzed by a UV or LED transilluminator or a digital gel imaging system.

  • Comes in a non-reducing format lacking DTT and is more sensitive than silver staining
  • Fluorescently pre-stains protein samples prior to gel electrophoresis
  • Visualize proteins directly within the gel using a UV or LED transilluminator
TRUSTED_SUSTAINABILITY

Specifications

Buffer Loading Buffer
Color Clear
Concentration 3X
Detection Method Electrophoresis
Gel Type Native-PAGE
Volume (Metric) 1 mL
Form Liquid
Quantity 1 Vial
Storage Requirements -20°C
Product Type Loading Buffer
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.