Learn More
Molecular Dimensions QUICK FOLD PROTEIN REFOLDING K
SDP
Supplier:  Molecular Dimensions MD128
Encompass
Protein Crystallography. The QuickFold Protein Refolding Kit enables researchers to pinpoint the critical factors for refolding their protein in as little as 1 hour. Includes buffer for 10 experiments, as well as supplemental Dithiothreitol and a Glutathione Redox System.
Catalog No. NC0071670
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.