Learn More
Perkin Elmer US LLC Quick Press Die Set - 7 mm
Supplier:  Perkin Elmer US LLC 01862289
Encompass_Preferred
The Quick Press provides a fast and economical way to prepare high quality sample pellets for FT-IR analysis. Pellets with diameters of 1, 3, or 7 mm can be prepared, depending on the die sets used. The required Pellet Holder for the Quick Press (51002593) is sold separately.
Catalog No. 50-211-2117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.