Learn More
Warner Instruments™ Quick Release Magnetic Imaging Chamber
Catalog No. BTX641946 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Compatibility:
12 mm Coverslip
Catalog No. Compatibility
BTX641946 12 mm Coverslip
1 options
Catalog No. BTX641946 Supplier Warner Instruments™ Supplier No. 641946
Low-profile quick release chamber supports rapid exchange of 12 mm round coverslips during successive experiments.
  • Forms glass coverslip-bottomed imaging chambers
  • Available in open- and closed-bath designs
  • Low-profile design enables low-angle electrode entry
  • Magnetic top and bottom assembly uses rare-earth magnets for QR Series models
  • Silicone O-ring seals against the glass coverslip
  • Compatible with 1 and 1.5 thickness coverslips
  • Anodized aluminum base supports heat transfer
Specifications
Compatibility 12 mm Coverslip