Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Quick-Release Pipet Pump PROMO

Catalog No. S23825S24 Shop All Eisco Products
Change view
Click to view available options
Volume (Metric):
2 mL
10 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S23825S24 2 mL
S23826S24 10 mL
S23827S24 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S23825S24 Supplier Eisco™ Supplier No. S23825S24

Quick-Release Pipet Pump is a pipetting device for precise pipetting and quick emptying.

  • Rotatable knurled thumb wheel to draw up or dispense liquids
  • Releaser to be pushed in for emptying
  • Molded chuck with inner silicon rubber collect to hold pipettes of standard sizes
  • Resists acids and alkalis
  • Can be disassembled for cleaning

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette
Grip Type Ergonomic
Material Polypropylene, Silicone Rubber
Number of Channels 1
Sterility Non-sterile
Type Pipette Pump
Volume (Metric) 2 mL
For Use With (Application) Precise Pipetting and Quick Emptying
Resistance Acids and Alkalis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.