Learn More
Thomas Scientific 0.2ML 8-STP W ATT FLAT CAPS, Nat
SDP
Supplier:  Thomas Scientific 1148A25
Encompass_Preferred
8 Quickstrip Q, 0.2ml PCR Tubes w/ Individual Flat Optical Caps, Certified RNase, DNase and Pyrogen free, 120 Strips/unit
Catalog No. NC1193406