Learn More
Agilent Technologies QUICKCHANGE II-MUTAGENESIS
Supplier:  Agilent Technologies 200524
Encompass
QuikChange II site-directed mutagenesis kit, Contains: PfuUltra High Fidelity DNA polymerase, dNTP mix, Dpn I restriction enzyme, QuikChange control plasmid and control primers, XL-1 supercompetent cells, pUC18 control plasmid
Catalog No. NC9053341
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.