Learn More
Zymo Research Corporation Sample Quick-RNA™ MicroPrep Kit (10 Preps) w/Zymo-Spin™ IC Columns (Capped)
SDP
Supplier:  Zymo Research Corporation R1050S
Encompass
Free Sample The Quick-RNA Microprep Kit is designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 106) and tissue samples (up to 5 mg). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17-200 nt) in about 10 minutes. The procedure is simple. Add the provided RNA Lysis Buffer to a sample, and then purify the RNA using the Zymo-Spin™Columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing etc. In addition, the kit can be used for the enrichment of small and large RNAs into separate fractions.
Catalog No. NC3345963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.