Learn More
Zymo Research Corporation Sample Quick-RNA™ MiniPrep Plus Kit (10 Preps) w/Zymo-Spin™ IIICG Columns (Capped) & Spin-Away™ Filters
SDP
Supplier:  Zymo Research Corporation R1057S
Encompass
Free Sample The Quick-RNA Miniprep Plus Kit is designed for the easy, reliable, and rapid isolation of DNA-free RNA from cells, all tissue types, whole blood, and biological fluids. The provided DNA/RNA Shield stabilizes samples, allowing them to be stored without the need for immediate freezing or processing for up to one month. Furthermore, DNA/RNA Shield™ inactivates RNases as well as microbial pathogens (viruses, bacteria, etc.). The procedure combines a unique buffer system with Zymo-Spin Column technology to yield high quality total RNA (including small RNAs 17-200 nt). Simply add DNA/RNA Shield and Proteinase K to extract total RNA from any tissue, then purify the RNA using the Zymo-Spin Column. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing, etc. In addition, the kit can be used for the enrichment of small and large RNAs in two separate fractions.
Catalog No. NC3345966
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.