Learn More
Description
- Crystal Violet
- Grams Iodine
- Safranin
- Decolorizer
Specifications
Specifications
| Quantity | 1 Kit |
| Includes | One bottle of each Crystal Violet, Grams Iodine, Safranin and Decolorizer. |
| For Use With (Application) | MGS-80, MGS-EW-100, MGS80-RC-9000, MGS80-TK-7000 |
| Composition | Weight: 37 lb.L x W x H: 38.1 x 38.1 x 41.6cm (15 X 15 X 16 in.) |
| Storage Requirements | Keep container tightly closed. Store in a cool, dry place |
| Stain Type | Crystal Violet, Grams Iodine, Safranin and Decolorizer. |
| Product Type | Reagent Stain Kit |
| Packaging | Screw cap bottles |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
