Learn More
United Scientific™ Quiet Air Source with Variable Output
SDPSCI_ED
Catalog No. S139508 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Equipment):
Air Track
Catalog No. For Use With (Equipment)
S139508 Air Track
1 options
This item is not returnable. View return policy
Catalog No. S139508 Supplier United Scientific™ Supplier No. QAS001V
Add to Cart
Add to Cart
This item is not returnable. View return policy
Air source for use with a wide range of air tracks.
  • This low noise (< 58 dB) air source is an ideal accessory for air tracks, air tables, and other pneumatic equipment in physics labs.
  • The 250 W motor produces a clean air output with a flow rate of 35 m3/hour at a 0.70 kPa overpressure.
  • Shipping weight: 12 lb.
  • Dimensions: 16 x 19 x 10 in.
Specifications
For Use With (Equipment) Air Track
Product Type Quiet Air Source
Prop 65 Warning: Cancer and reproductive harm