Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ QUIK-ALIGN™ Gothic Permanent Adhesive Vinyl Numbers, Letters & Special Characters Label Kit - 4 in H, Red

Catalog No. 19108137 Shop All Brady Corporation Products
Change view
Click to view available options
QUIK-ALIGN™ Gothic Permanent Adhesive Vinyl Numbers, Letters & Special Characters Label Kit - 4 in H, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-108-137 QUIK-ALIGN™ Gothic Permanent Adhesive Vinyl Numbers, Letters & Special Characters Label Kit - 4 in H, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-108-137 Supplier Brady™ Supplier No. 52217

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Quik-Align™ die-cut numbers and letters are self-spacing and self-aligning: the characters interlock to ensure perfect spacing for professional-looking results every time.

  • Traditional sans serif Gothic font provides excellent legibility even at a distance
  • Card-mounted vinyl film characters features a pressure sensitive adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 2-4 years

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 2979
Description Label
Adhesive Type Permanent Acrylic
Color Red
Format Die-cut
Material Vinyl
Product Type Label
No. per Pack 1 Kit
Quantity Kit
Size (English) 4.25 in. H
Length (English) 4.25 in.
Height (English) Character 4 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.