Learn More
Brady™ Quik-Align™ Gothic Style Kits, White
Catalog No. 19108134 Shop All Brady Corporation Products
Change view
Click to view available options
Height (English):
1 in.
2 in.
3 in.
4 in.
6 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
19-108-134 2 in.
19-108-132 1 in.
19-108-135 3 in.
19-108-136 4 in.
19-108-138 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-108-134 Supplier Brady™ Supplier No. 52208
For identification of equipment, bins and areas of the warehouse required by facility safety and maintenance. Brady Quik-Align Gothic Style Kits are made of vinyl and can be used to mark tool cribs, bins, shelves, poles, panels, switches and equipment.
  • Self-spacing, self-aligning diecuts
  • Made of B-933 vinyl
  • For general purpose indoor or outdoor marking
Specifications
Product Type Gothic Quik-Align Kit
Color White
Material Vinyl (B-933)
Height (English) 2 in.
Thickness (English) 0.0049 in.
Adhesive Type Permanent
Description Abrasion, chemical resistant
Format Die-cut