Learn More
AdipoGen Quinacrine mustard 2HCl
SDP
Supplier:  AdipoGen CDXQ0011M025
Encompass_Preferred
Chemical. CAS 4213-45-0. Formula C23H28Cl3N3O . 2HCl. MW 468.9 . 72.9. Synthetic. Fluorescent probe for labeling chromosomal DNA. Also used in measurement of sequence specificity with Taq DNA polymerase.
Catalog No. 50-256-0163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.