Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldine, 96%

Catalog No. AAA1341222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1341222 100 g
AAA1341214 25 g
AAA1341236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1341222 Supplier Thermo Scientific Chemicals Supplier No. A1341222
Add to Cart

CAS: 91-63-4 | C10H9N | 143.19 g/mol

It is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.

Solubility
Soluble in water (2.5g/L).

Notes
Air and Light Sensitive. Store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides. It may discolor to brown upon storage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-63-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006756
InChI Key SMUQFGGVLNAIOZ-UHFFFAOYSA-N
Synonym quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech
PubChem CID 7060
IUPAC Name 2-methylquinoline
SMILES CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point -2°C
Density 1.058
Boiling Point 247°C to 248°C
Flash Point 79°C (174°F)
Odor Odorless
Refractive Index 1.612
Quantity 100 g
Beilstein 110309
Sensitivity Air and light sensitive
Merck Index 14,8047
Solubility Information Soluble in water (2.5g/L).
Formula Weight 143.19
Percent Purity ≥97%
Chemical Name or Material Quinaldine
Show More Show Less
Hazard Category H227-H302+H312-H315-H319-H335-H341
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P374-P501c
EINECSNumber 202-085-1
RTECSNumber UZ9625000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.