Learn More
Selleck Chemical LLC Quinestrol S3671-25mg
SDP
Supplier:  Selleck Chemical LLC S367125MG
Encompass_Preferred
Quinestrol (Ethinylestradiol 3-cyclopentyl ether EE2CPE W-3566) is a synthetic steroidal estrogen which is used in hormone replacement therapy and occasionally to treat breast cancer and prostate cancer
Catalog No. 50-311-0727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.