Learn More
Apexbio Technology LLC VPC 23019 449173-19-7 5mg
SDP
Supplier:  Apexbio Technology LLC B76115
Encompass_Preferred
VPC 23019 (CAS 449173-19-7) is a small molecule antagonist targeting sphingosine-1-phosphate receptors S1P1 and S1P3 It disrupts S1P-mediated signaling with an IC50 of 8 nM for S1P1 inhibition VPC 23019 abolishes S1P-induced cellular migration Gi-dependent Rac activation and tube formation in relevant cell models In vascular endothelial cells it does not alter basal nitric oxide production but attenuates Angiotensin II-stimulated NO release The compound also suppresses S1P-induced transendothelial migration and partially inhibits S1P-evoked calcium elevation in fibroblasts In vascular reactivity assays VPC 23019 modulates S1P-induced contraction in endothelium-intact vessels without affecting responses to other vasoactive agents VPC 23019 supports research into S1P receptor functions in cellular signaling and vascular physiology
Catalog No. 50-000-91716
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.