Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine, 98+%, anhydrous

Catalog No. AC163702500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163702500 250 g Glass bottle
AC163700100 10 g Glass bottle
AC163700500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163702500 Supplier Thermo Scientific Chemicals Supplier No. 163702500
Add to Cart

CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00198096
InChI Key LOUPRKONTZGTKE-ZCXAXGJJNA-N
Synonym 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol
PubChem CID 129316724
IUPAC Name (5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl)-[6-(trideuteriomethoxy)quinolin-4-yl]methanol
SMILES [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 176.0°C to 177.0°C
Color White
Loss on Drying 1% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 g
Fieser 06,501; 07,311; 08,340; 09,403; 10,338; 11,456
Merck Index 15, 8177
Specific Rotation Condition − 172.00 (20.00°C c=1,C2H5OH)
Specific Rotation − 172.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkalies and acids (formation of salts),soluble in alcohol,ether,chloroform,glycerol,and carbon disulfide
Formula Weight 324.42
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Quinine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s
EINECSNumber 205-003-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.