Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinine hydrochloride, Tocris Bioscience™
SDP

Catalog No. 411450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-145-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-145-0 Supplier R&D Systems Supplier No. 4114/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antimalarial

Quinine hydrochloride displays antimalarial and muscle relaxant properties. Stereoisomer of quinidine (Cat. No. 4108); closely related to chloroquine (Cat. No. 4109).

Chemical Identifiers

CAS 130-89-2
Molecular Formula C20H25ClN2O2
Molecular Weight (g/mol) 360.88
MDL Number MFCD00078498
InChI Key PKLFKSABAIAQBR-UHFFFAOYNA-N
Synonym quinine hydrochloride, s-2r,4r,5s-5-ethenyl-1-azabicyclo 2.2.2 octan-2-yl 6-methoxyquinolin-4-yl methanol hydrochloride
PubChem CID 134129329
IUPAC Name hydrogen {4-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol chloride
SMILES [H+].[Cl-].COC1=CC=C2N=CC=C(C(O)C3CC4(CCN3CC4)C=C)C2=C1

Specifications

Quantity 50 mg
Formula Weight 360.88
Percent Purity >98%
Chemical Name or Material Quinine hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.