Learn More
AnaSpec QUININE SULFATE FLUORESCENCE
Supplier:  AnaSpec AS80040
Encompass_Preferred
Quinine sulfate Dihydrate *Fluorescence Reference Standard, ; MW 782.96; (100 mg) Reference standard for measuring fluorescence quantum yield
Catalog No. 50-850-220
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.