Learn More
Abcam Quinol 1h, Antitumor Agent, Solid, 25 mg
Description
- Purity: >99%
- Molecular Weight: 365.4 Da
- Form: Solid
- Activity: Potent, selective antitumor agent. Antitumor activity observed in HCT 116 (GI50 = 40 nM) and HT 29 (GI50 = 380 nM), and in breast cancer cell lines. Potent inhibitory effect in NCI 60 cells (GI50 = 16 nM and LC50 = 2.24 μM). Active in vitro.
- Storage: Store at -20°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Quinol 1h |
| Shipping Condition | Ambient |
| Form | Solid |
| Alternate Part Number | AB144639 |
| Buffer | 100% Quinol 1h |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Quinol 1h |
| Quantity | 25 mg |
| For Use With (Application) | Antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.