Learn More
Abcam Quinol 1h, Antitumor agent, 5MG
Supplier:  Abcam AB1446395MG
Encompass_Preferred
MW 365.4 Da, Purity >99%. Potent, selective antitumor agent. Antitumor activity observed in HCT 116 (GI₅₀ = 40 nM) and HT 29 (GI₅₀ = 380 nM), and in breast cancer cell lines. Potent inhibitory effect in NCI 60 cells (GI₅₀ =16 nM and LC₅₀ = 2.24 μM). Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5539