Learn More
Abcam Quinol 2, Antitumor Agent, >99% Purity, Solid, 25 mg
Description
- Molecular weight: 243.28 Da
- Purity: >99%
- Form: Solid
- Activity: Selective antitumor agent active against colon, renal and breast carcinomas in vitro and ex vivo.
- Mechanism: Inhibits thioredoxin reductase.
- Potency: GI50 = 0.23 μM and LC50 = 3.39 μM in NCI 60 cells.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Buffer | 100% Quinol 2 |
| Target Molecule | Thioredoxin Reductase |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Formulation | 100% Quinol 2 |
| Quantity | 25 mg |
| For Use With (Application) | Cancer research, inhibition of thioredoxin reductase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.