Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Quinol 2, Antitumor Agent, >99% Purity, Solid, 5 mg

Catalog No. 502705537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5537 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5537 Supplier Abcam Supplier No. AB1446405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective antitumor agent that inhibits thioredoxin reductase.

  • Purity: >99%
  • Molecular Weight: 243.28 Da
  • Form: Solid
  • Activity: Selective antitumor agent; active against colon, renal and breast carcinomas in vitro and ex vivo
  • Mechanism: Inhibits thioredoxin reductase with potencies correlated with antiproliferative and cytotoxic effects
  • Potency: Potent inhibitory effect in NCI 60 cells (GI50 =0.23 μM and LC50 = 3.39 μM)
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Thioredoxin Reductase
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Quantity 5 mg
For Use With (Application) Antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.