Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline-2-carboxaldehyde, 97%

Catalog No. AAL2020206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2020206 5 g
AAL2020214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2020206 Supplier Thermo Scientific Chemicals Supplier No. L2020206
Add to Cart

CAS: 5470-96-2 | C10H7NO | 157.17 g/mol

2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-96-2
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00075032
InChI Key WPYJKGWLDJECQD-UHFFFAOYSA-N
Synonym 2-quinolinecarboxaldehyde, quinoline-2-carboxaldehyde, 2-quinolinecarbaldehyde, 2-formylquinoline, quinaldaldehyde, quinolinecarboxaldehyde, quinoline aldehyde, quinoline-2-aldehyde, zlchem 599, pubchem2277
PubChem CID 79619
IUPAC Name quinoline-2-carbaldehyde
SMILES O=CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Beilstein 113061
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material Quinoline-2-carboxaldehyde
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-804-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.