Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline N-Oxide Hydrate 97.0+%, TCI America™
SDP

Catalog No. Q007225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q007225G 25 g
Q00725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q007225G Supplier TCI America Supplier No. Q007225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 198878-42-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00149472
InChI Key GIIWGCBLYNDKBO-UHFFFAOYSA-N
PubChem CID 152363
IUPAC Name quinolin-1-ium-1-olate
SMILES [O-][N+]1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 59°C
Boiling Point 173°C
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinoline N-Oxide Hydrate
RTECSNumber VC2335000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.