Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinuclidine, 97+%

Catalog No. p-7036237
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54498MD 250 mg
AAH5449803 1 g
AAH5449806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54498MD Supplier Thermo Scientific Chemicals Supplier No. H54498MD
Only null left
Add to Cart

CAS: 100-76-5 | C7H13N | 111.188 g/mol

Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-76-5
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00006690
InChI Key SBYHFKPVCBCYGV-UHFFFAOYSA-N
Synonym quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi
PubChem CID 7527
ChEBI CHEBI:38420
IUPAC Name 1-azabicyclo[2.2.2]octane
SMILES C1CN2CCC1CC2

Specifications

Melting Point 157°C to 160°C
Quantity 250 mg
UN Number UN2811
Beilstein 103111
Sensitivity Air Sensitive
Merck Index 14,8081
Solubility Information Soluble in alcohol,diethyl ether,water and organic solvents.
Formula Weight 111.19
Percent Purity ≥97%
Chemical Name or Material Quinuclidine
Hazard Category H301-H310-H314-H335
Hazard Statement GHS H Statement
H301-H310-H318-H315
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-887-1
RTECSNumber CL5594625
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.