Learn More
Abcam Quisqualic Acid, Group I mGlu Agonist, Solid, 20 mM/mL
Description
- Molecular Weight: 189.13 Da
- Purity: >99%
- Potency: Potent group I mGlu agonist and AMPA receptor agonist
- Form: Solid
- Storage Buffer: 100% Quisqualic acid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | mGlu Receptor, AMPA Receptor |
| Buffer | 100% Quisqualic acid |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% Quisqualic acid |
| Quantity | 20 mM/mL |
| For Use With (Application) | Group I mGlu agonist, AMPA receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.