Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam QX-314 Bromide, Sodium Channel Blocker, 343.3 Da, 100 mg
Description
- Molecular weight: 343.3 Da
- Purity: >99%
- Description: Membrane-impermeable quaternary lidocaine derivative
- Mechanism: Blocks voltage-sensitive Na+ conductance when applied intracellularly
- Form: Solid
- Solubility: Soluble in 1 mL water to give specified mM/mL concentration
- Storage buffer: 100% Lidocaine N-ethyl bromide
- Citation count: 14
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at 4°C. |
| Buffer | 100% Lidocaine N-ethyl bromide |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Target Molecule | Na+ Channels |
| Formulation | 100% Lidocaine N-ethyl bromide |
| Quantity | 100 mg |
| For Use With (Application) | Na+ channel blocking |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?