Learn More
Abcam QX-314 Bromide, Sodium Channel Blocker, 343.3 Da, 250 mg
Description
- Molecular Weight: 343.3 Da
- Purity: >99%
- Form: Solid
- Description: Membrane impermeable quaternary lidocaine derivative that blocks voltage-sensitive Na+ conductance when applied intracellularly.
- Storage: Store at +4°C.
- Shipping: Ambient - Can Ship with Ice
- Citations: 14
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Na+ Channels |
| Buffer | 100% Lidocaine N-ethyl bromide |
| Form | Solid |
| Formulation | 100% Lidocaine N-ethyl bromide |
| Quantity | 250 mg |
| For Use With (Application) | Na+ channel blocking |
| Storage Requirements | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.